Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 10 | 1013 | ||
NM_001282644.1 | NP_001269573.1 | 10 | 1011 | |||
NM_001282645.1 | NP_001269574.1 | 9 | 952 | |||
NM_001323265.1 | NP_001310194.1 | 10 | 1068 | |||
NM_001323266.1 | NP_001310195.1 | 9 | 950 | |||
NM_001323267.1 | NP_001310196.1 | 10 | 1011 | |||
NM_006257.4 | NP_006248.1 | 10 | 1013 | |||
XM_005252496.4 | XP_005252553.1 | 10 | 1113 | |||
XM_005252497.4 | XP_005252554.1 | 10 | 1115 | |||
XM_006717465.3 | XP_006717528.1 | 12 | 3508 | |||
XM_017016410.1 | XP_016871899.1 | 11 | 3419 | |||
XM_017016411.1 | XP_016871900.1 | 11 | 3447 | |||
GenBank mRNA | AK293935.1 | - | 9 | 890 | ||
AK308099.1 | - | 10 | 986 | |||
AK312411.1 | - | 10 | 963 | |||
AY702977.1 | - | 9 | 913 | |||
BC101465.1 | - | 9 | 932 | 8068971 | ||
BC113359.1 | - | 9 | 932 | 8322411 | ||
BX647657.1 | - | 9 | 1098 | |||
L01087.1 | - | 10 | 1007 | |||
L07032.1 | - | 9 | 934 |