Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002754.4 | NP_002745.1 | 11, 12 | 1127 | ||
NR_072996.1 | 9, 10 | 975 | ||||
GenBank mRNA | AB209586.1 | - | 8, 9 | 1933 | ||
AB451221.1 | - | 12, 13 | 1028 | |||
AB451344.1 | - | 12, 13 | 1028 | |||
AF004709.1 | - | 11, 12 | 1076 | |||
AF015256.1 | - | 11, 12 | 1088 | |||
AF092535.1 | - | 11, 12 | 1049 | |||
AF100546.1 | - | 11, 12 | 1030 | |||
AK222922.1 | - | 9, 10 | 854 | |||
BC000433.2 | - | 11, 12 | 1069 | 2819932 | ||
BC001641.2 | - | 11, 12 | 1069 | 2819932 | ||
BC004428.1 | - | 11, 12 | 1069 | 2819932 | ||
BC085196.1 | - | 9, 10 | 945 | 6107307 | ||
BT007221.1 | - | 11, 12 | 1028 | |||
CR536490.1 | - | 11, 12 | 1028 | |||
KU178281.1 | - | 11, 12 | 1028 | |||
KU178282.1 | - | 11, 12 | 1061 | |||
U93232.1 | - | 11, 12 | 1035 | |||
Y10488.1 | - | 11, 12 | 1050 |