Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006260.4 | NP_006251.1 | 4, 5 | 516 | ||
XM_017020674.1 | XP_016876163.1 | 4, 5 | 451 | |||
XM_017020675.1 | XP_016876164.1 | 1, 3 | 1024 | |||
GenBank mRNA | AK292947.1 | - | 4, 5 | 516 | ||
BC047936.1 | - | 4, 5 | 498 | 5215035 |