Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000684.2 | NP_000675.1 | 1 | 774 | ||
GenBank mRNA | J03019.1 | - | 1 | 774 |
Set Membership: |
G-protein coupled receptor |