Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146528.2 | NP_666739.1 | 2 | 819 | ||
GenBank mRNA | BC120862.1 | - | 1 | 650 | 40129785 | |
BC137831.1 | - | 1 | 650 | 8860850 |