Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002772.2 | NP_002763.2 | 13 | 1515 | ||
XM_011529654.1 | XP_011527956.1 | 15 | 1789 | |||
XM_011529655.1 | XP_011527957.1 | 16 | 1707 | |||
XM_011529656.2 | XP_011527958.1 | 16 | 1714 | |||
XM_011529657.1 | XP_011527959.1 | 14 | 1744 | |||
XM_011529658.1 | XP_011527960.1 | 13 | 1708 | |||
XM_011529659.1 | XP_011527961.1 | 14 | 1679 | |||
GenBank mRNA | BC111749.1 | - | 13 | 1504 | 40008569 | |
U09860.1 | - | 13 | 1524 |