Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146952.1 | NP_667163.1 | 1 | 815 | ||
GenBank mRNA | BC120696.1 | - | 1 | 881 | 40129619 |