Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146965.1 | NP_667176.1 | 1 | 619 | ||
GenBank mRNA | BC125323.1 | - | 1 | 713 | 40129838 | |
BC125349.1 | - | 1 | 713 | 40129864 |