Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147016.2 | NP_667227.2 | 1 | 618 |
Gene Symbol: Olfr1051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207562.1 | NP_997445.1 | 1 | 618 |