Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153068.3 | NP_694708.2 | 4 | 758 | ||
GenBank mRNA | AK029240.1 | - | 4 | 702 | ||
AK031203.1 | - | 4 | 760 | |||
AK046566.1 | - | 4 | 700 | |||
AK048356.1 | - | 4 | 703 | |||
AK050243.1 | - | 4 | 738 | |||
AK084455.1 | - | 4 | 701 | |||
AK085016.1 | - | 4 | 701 | |||
AY531389.1 | - | 4 | 697 | |||
BC027084.1 | - | 5 | 405 | 5356224 | ||
BC031731.1 | - | 623 | ||||
BC113161.1 | - | 5 | 716 | 40060184 |