Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153680.2 | NP_710147.1 | 5 | 478 | ||
GenBank mRNA | AK047265.1 | - | 5 | 478 | ||
AK077650.1 | - | 5 | 480 | |||
AK077780.1 | - | 5 | 480 | |||
AK156299.1 | - | 5 | 529 | |||
AK219724.1 | - | 2 | 123 | |||
BC023732.1 | - | 5 | 494 | 5352355 | ||
BC026571.1 | - | 5 | 478 | 4986268 | ||
BC029675.1 | - | 5 | 465 |