Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002810.2 | NP_002801.1 | 2 | 157 | ||
XM_005245354.2 | XP_005245411.1 | 2 | 178 | |||
GenBank mRNA | AB033605.1 | - | 2 | 109 | ||
BC002365.2 | - | 2 | 107 | 2820813 | ||
BC072008.1 | - | 2 | 121 | 6285035 | ||
U24704.1 | - | 2 | 157 |