Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174852.3 | NP_777277.2 | 7, 8 | 1635 | ||
GenBank mRNA | AK050751.1 | - | 1, 2 | 51 | ||
AK089226.1 | - | 7, 8 | 1698 | |||
AK129383.1 | - | 4, 5 | 782 | |||
AK141796.1 | - | 7, 8 | 1539 | |||
AK155354.1 | - | 7, 8 | 1676 | |||
AK170155.1 | - | 7, 8 | 1723 | |||
AK170187.1 | - | 6, 7 | 930 | |||
BC043080.1 | - | 7, 8 | 1238 | 6400617 |