Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178744.2 | NP_848859.1 | 2 | 1556 | ||
GenBank mRNA | AK029762.1 | - | 2 | 1304 | ||
AK035036.1 | - | 2 | 1636 | |||
AK049397.1 | - | 2 | 1556 | |||
AK131985.1 | - | 2 | 1239 | |||
AY611626.1 | - | 2 | 1556 | |||
BC012239.1 | - | 2 | 1220 | 4037044 |