Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201354.2 | NP_958742.2 | 6 | 809 | ||
GenBank mRNA | BC058104.1 | - | 6 | 667 | 6833739 |