Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfasc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160316.1 | NP_001153788.1 | 8 | 960 | ||
NM_001160317.1 | NP_001153789.1 | 10 | 1196 | |||
NM_001160318.1 | NP_001153790.1 | 9 | 1246 | |||
NM_182716.4 | NP_874385.1 | 10 | 1163 | |||
GenBank mRNA | AJ543322.1 | - | 10 | 1132 | ||
AK047548.1 | - | 2 | 3020 | |||
AK052440.1 | - | 8 | 1036 | |||
AK129207.1 | - | 9 | 1246 | |||
AK138636.1 | - | 10 | 1111 |
Set Membership: |
G-protein coupled receptor |