Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199310.2 | NP_955014.2 | 3, 4 | 488 | ||
GenBank mRNA | AK132604.1 | - | 3, 4 | 480 | ||
AK133813.1 | - | 3, 4 | 462 | |||
AK147152.1 | - | 3, 4 | 464 | |||
BC023926.1 | - | 3, 4 | 482 | 5324154 | ||
BC052687.1 | - | 3, 4 | 455 |