Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172980.2 | NP_766568.1 | 15 | 2834 | ||
GenBank mRNA | AF079853.2 | - | 14 | 1709 | ||
AK080957.1 | - | 15 | 2835 | |||
AK152776.1 | - | 14 | 1769 | |||
BC055376.1 | - | 14 | 1728 | 30077988 |