Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081162.1 | NP_001074631.1 | 13 | 1604 | ||
GenBank mRNA | AK052665.1 | - | 8 | 2473 | ||
AK158115.1 | - | 13 | 1558 | |||
BC036154.1 | - | 3 | 1182 | |||
BC111884.1 | - | 12 | 1399 | 40060299 |