Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030007.1 | NP_001025178.1 | 8 | 1020 | ||
NM_001128.5 | NP_001119.3 | 7 | 1011 | |||
GenBank mRNA | AB015317.1 | - | 7 | 716 | ||
AK128078.1 | - | 10 | 1566 | |||
AK225906.1 | - | 8 | 779 | |||
AK293233.1 | - | 8 | 670 | |||
AK294546.1 | - | 8 | 864 | |||
AK299630.1 | - | 9 | 954 | |||
AK314484.1 | - | 7 | 1016 | |||
BC036283.1 | - | 8 | 985 | 4158263 | ||
BU902252.1 | - | 7 | 1009 | |||
Y12226.1 | - | 8 | 734 |