Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130184.1 | NP_001123656.1 | 6 | 661 | ||
NM_001130185.1 | NP_001123657.1 | 6 | 741 | |||
NM_001130186.1 | NP_001123658.1 | 6 | 699 | |||
NM_172303.4 | NP_758507.3 | 6 | 759 | |||
GenBank mRNA | AK049332.1 | - | 6 | 664 | ||
AK129445.1 | - | 6 | 654 | |||
AK147466.1 | - | 6 | 621 | |||
AK193039.1 | - | 1 | 56 | |||
BC020316.2 | - | 6 | 681 | 5250231 |