Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030007.1 | NP_001025178.1 | 20 | 2227 | ||
NM_001128.5 | NP_001119.3 | 19 | 2218 | |||
GenBank mRNA | AB015317.1 | - | 19 | 1923 | ||
AK055122.1 | - | 3 | 838 | |||
AK128078.1 | - | 22 | 2773 | |||
AK225906.1 | - | 20 | 1986 | |||
AK314484.1 | - | 19 | 2223 | |||
AL110198.1 | - | 3 | 292 | |||
BC036283.1 | - | 20 | 2192 | 4158263 | ||
Y12226.1 | - | 20 | 1941 |