Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177769.4 | NP_808437.2 | 6 | 691 | ||
GenBank mRNA | AK029207.1 | - | 6 | 572 | ||
AK132239.1 | - | 6 | 692 | |||
BC049160.1 | - | 6 | 554 | |||
BC120566.1 | - | 5 | 407 | |||
BC144961.1 | - | 5 | 407 | |||
BC144963.1 | - | 5 | 369 | 9053511 |