Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177772.4 | NP_808440.2 | 10 | 1013 | ||
GenBank mRNA | AK028764.1 | - | 10 | 1010 | ||
AK132518.1 | - | 10 | 1049 | |||
BC112376.1 | - | 10 | 1001 | 40057331 |