siRNA Details
Gene Symbol: Adamts16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172053.2 | NP_742050.2 | 14 | 2386 | ||
GenBank mRNA | AK031314.1 | - | 14 | 2343 | ||
AK173338.1 | - | 14 | 2322 | |||
BC034739.1 | - | 14 | 2399 | 4951547 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
