Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178856.1 | NP_849187.1 | 4 | 399 | ||
GenBank mRNA | AK014776.1 | - | 4 | 413 | ||
AK028375.1 | - | 4 | 351 | |||
AK030758.1 | - | 416 | ||||
BC082565.1 | - | 4 | 399 | 30551463 |