Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duxbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183389.1 | NP_899245.1 | 5 | 1229 | ||
GenBank mRNA | AK141686.1 | - | 5 | 905 | ||
BC055782.1 | - | 5 | 1229 | 6403934 | ||
EF472598.1 | - | 5 | 966 |
Gene Symbol: Duxbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177538.1 | NP_001171009.1 | 5 | 1219 |
Gene Symbol: Duxbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177539.1 | NP_001171010.1 | 1219 |