Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172126.2 | NP_742124.2 | 1 | 2215 | ||
GenBank mRNA | AK132801.1 | - | 1 | 2102 | ||
AK132804.1 | - | 1 | 2106 | |||
AK133150.1 | - | 1 | 2116 | |||
AK133286.1 | - | 1 | 2106 | |||
BC052093.1 | - | 1 | 2215 | 5695131 | ||
U22056.1 | - | 1 | 1177 |