Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175034.3 | NP_778199.2 | 9 | 1504 | ||
GenBank mRNA | AB085629.1 | - | 9 | 1507 | ||
AK089225.1 | - | 9 | 1508 | |||
AY236527.1 | - | 9 | 1474 | |||
BC094232.1 | - | 9 | 1464 | 30538386 | ||
BC144866.1 | - | 9 | 1499 | 9053414 | ||
BC145643.1 | - | 9 | 1499 | 40130946 |