Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256516.1 | NP_001243445.1 | 4 | 1461 | ||
NM_001256517.1 | NP_001243446.1 | 4 | 1430 | |||
NM_001256518.1 | NP_001243447.1 | 5 | 1712 | |||
NM_001256519.1 | NP_001243448.1 | 6 | 1641 | |||
NM_001256520.1 | NP_001243449.1 | 3 | 1332 | |||
NM_178761.5 | NP_848876.2 | 3 | 1300 | |||
GenBank mRNA | AK029711.1 | - | 5 | 1717 | ||
AK035113.1 | - | 3 | 1279 | |||
AK085166.1 | - | 4 | 1390 | |||
BC003258.1 | - | 1 | 1103 | 3500812 | ||
BC034706.1 | - | 4 | 1408 | 3582315 |