Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176996.4 | NP_795970.3 | 6 | 1697 | ||
GenBank mRNA | AK193396.1 | - | 2 | 84 | ||
AK194911.1 | - | 2 | 89 | |||
BC048091.1 | - | 6 | 1503 | 6486595 | ||
BC096028.1 | - | 6 | 1216 | 6850065 | ||
CA749812.1 | - | 3 | 297 | |||
FJ883552.1 | - | 6 | 1206 |
Set Membership: |
G-protein coupled receptor |