Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183029.2 | NP_898850.2 | 9 | 1336 | ||
GenBank mRNA | AK044984.1 | - | 8 | 852 | ||
AK049196.1 | - | 9 | 1033 | |||
AK077404.1 | - | 9 | 1017 | |||
AK159654.1 | - | 9 | 1069 | |||
AK170531.1 | - | 8 | 840 | |||
AK190144.1 | - | 2 | 204 | |||
AK194213.1 | - | 2 | 198 | |||
AY531659.1 | - | 9 | 1000 | |||
BC023758.1 | - | 9 | 1034 | |||
BC054552.1 | - | 9 | 1056 |