Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11540
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
20
3030
NM_001171025.1
NP_001164496.1
19
3039
NM_002839.3
NP_002830.1
36
4900
NM_130391.3
NP_569075.2
21
3039
NM_130392.3
NP_569076.2
21
3042
NM_130393.3
NP_569077.2
19
3012
XM_006716817.3
XP_006716880.1
37
5249
XM_006716823.2
XP_006716886.1
35
5195
XM_006716825.3
XP_006716888.1
35
5194
XM_006716827.3
XP_006716890.1
32
5134
XM_006716832.3
XP_006716895.1
29
3965
XM_006716833.3
XP_006716896.1
31
3967
XM_006716834.3
XP_006716897.1
29
3968
XM_006716835.3
XP_006716898.1
30
3958
XM_006716837.3
XP_006716900.1
29
3938
XM_006716838.3
XP_006716901.1
29
3937
XM_006716839.3
XP_006716902.1
28
3923
XM_011517992.2
XP_011516294.1
35
5191
XM_017014958.1
XP_016870447.1
37
5257
XM_017014959.1
XP_016870448.1
38
5329
XM_017014960.1
XP_016870449.1
36
5218
XM_017014961.1
XP_016870450.1
36
5202
XM_017014962.1
XP_016870451.1
28
4390
XM_017014963.1
XP_016870452.1
37
5254
XM_017014964.1
XP_016870453.1
37
5253
XM_017014965.1
XP_016870454.1
37
5256
XM_017014966.1
XP_016870455.1
36
5248
XM_017014967.1
XP_016870456.1
36
5243
XM_017014968.1
XP_016870457.1
36
5239
XM_017014969.1
XP_016870458.1
36
5239
XM_017014970.1
XP_016870459.1
36
5239
XM_017014971.1
XP_016870460.1
35
5228
XM_017014972.1
XP_016870461.1
36
5230
XM_017014973.1
XP_016870462.1
35
5224
XM_017014974.1
XP_016870463.1
36
5227
XM_017014975.1
XP_016870464.1
34
5213
XM_017014976.1
XP_016870465.1
36
5212
XM_017014977.1
XP_016870466.1
35
5213
XM_017014978.1
XP_016870467.1
36
5209
XM_017014979.1
XP_016870468.1
36
5208
XM_017014980.1
XP_016870469.1
34
5164
XM_017014981.1
XP_016870470.1
31
5120
XM_017014982.1
XP_016870471.1
32
4020
XM_017014983.1
XP_016870472.1
32
4017
XM_017014984.1
XP_016870473.1
31
4008
XM_017014985.1
XP_016870474.1
31
4002
XM_017014986.1
XP_016870475.1
31
3998
XM_017014987.1
XP_016870476.1
30
3983
XM_017014988.1
XP_016870477.1
30
3980
XM_017014989.1
XP_016870478.1
31
3974
XM_017014990.1
XP_016870479.1
31
3970
XM_017014991.1
XP_016870480.1
31
3970
XM_017014992.1
XP_016870481.1
30
3961
XM_017014993.1
XP_016870482.1
30
3956
XM_017014994.1
XP_016870483.1
30
3953
XM_017014995.1
XP_016870484.1
30
3959
GenBank mRNA
AB208869.1
-
19
2856
AB211400.1
-
35
4868
AK296392.1
-
4
297
BC106713.1
-
19
2987
40027581
BC106714.1
-
21
2990
40027582
BC106715.2
-
21
2987
40027584
BC106716.1
-
19
2960
40027585
L38929.1
-
26
4310
X54133.1
-
16
2990
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0