Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 12 | 1255 | ||
NM_001316677.1 | NP_001303606.1 | 10 | 928 | |||
NM_001323354.1 | NP_001310283.1 | 10 | 2648 | |||
NM_001323355.1 | NP_001310284.1 | 10 | 1061 | |||
NM_001323356.1 | NP_001310285.1 | 10 | 1055 | |||
NM_001323357.1 | NP_001310286.1 | 7 | 640 | |||
NM_006504.5 | NP_006495.1 | 11 | 1084 | |||
NM_130435.4 | NP_569119.1 | 8 | 731 | |||
XM_005252691.2 | XP_005252748.1 | 10 | 928 | |||
XM_011539994.2 | XP_011538296.1 | 10 | 1046 | |||
XM_011539995.1 | XP_011538297.1 | 10 | 1046 | |||
XM_011539996.1 | XP_011538298.1 | 10 | 1046 | |||
XM_011539998.1 | XP_011538300.1 | 10 | 1046 | |||
XM_017016467.1 | XP_016871956.1 | 10 | 1046 | |||
XM_017016468.1 | XP_016871957.1 | 10 | 835 | |||
XM_017016469.1 | XP_016871958.1 | 10 | 1046 | |||
GenBank mRNA | AJ315969.1 | - | 8 | 640 | ||
AJ430580.1 | - | 8 | 649 | |||
AK290414.1 | - | 8 | 667 | |||
AK291828.1 | - | 10 | 928 | |||
AK299498.1 | - | 9 | 806 | |||
AK300613.1 | - | 9 | 821 | |||
BC050062.1 | - | 12 | 1163 | 5274802 | ||
BX648180.1 | - | 7 | 646 | |||
X54134.1 | - | 11 | 856 |