Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177069.3 | NP_796043.1 | 6 | 713 | ||
GenBank mRNA | AK087710.1 | - | 6 | 714 | ||
AK087902.1 | - | 6 | 712 | |||
AK135949.1 | - | 6 | 714 | |||
AK136136.1 | - | 6 | 710 | |||
BC099397.1 | - | 6 | 695 | 30936381 |