Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285806.1 | NP_001272735.1 | 5 | 525 | ||
NM_177078.4 | NP_796052.2 | 5 | 486 | |||
GenBank mRNA | AK048763.1 | - | 5 | 451 | ||
AK076533.1 | - | 5 | 384 | |||
AK134672.1 | - | 5 | 463 | |||
AK141186.1 | - | 5 | 504 | |||
AK172513.1 | - | 5 | 438 |