Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166685.1 | NP_001160157.1 | 15 | 1883 | ||
NM_177088.3 | NP_796062.2 | 16 | 2012 | |||
GenBank mRNA | AK076400.1 | - | 16 | 1990 | ||
BC090675.1 | - | 16 | 1968 | |||
BC116308.1 | - | 15 | 1849 | 40095242 | ||
BC116309.1 | - | 15 | 1866 |