Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177190.5 | NP_796164.1 | 2 | 340 | ||
GenBank mRNA | AK082024.1 | - | 2 | 342 | ||
BC116926.1 | - | 1 | 161 | 40126245 | ||
BC116932.1 | - | 1 | 161 | 40126251 |