Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cass4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080820.2 | NP_001074289.1 | 5 | 1159 | ||
NM_001276422.1 | NP_001263351.1 | 5 | 1165 | |||
NM_001276423.1 | NP_001263352.1 | 5 | 1159 | |||
GenBank mRNA | AK036419.1 | - | 5 | 1133 | ||
AK080960.1 | - | 5 | 1133 | |||
AK089450.1 | - | 5 | 1160 | |||
AK170726.1 | - | 5 | 1159 | |||
BC117858.1 | - | 5 | 1053 | 40095182 | ||
BC117859.1 | - | 5 | 1052 | 40095183 | ||
BC129975.1 | - | 5 | 1050 | 40095184 | ||
BC129976.1 | - | 5 | 1058 | 40095185 |