Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177225.4 | NP_796199.1 | 2 | 199 | ||
GenBank mRNA | AK032588.1 | - | 2 | 167 | ||
AK044142.1 | - | 2 | 200 | |||
AK080536.1 | - | 2 | 167 | |||
BC119383.1 | - | 2 | 200 | 8734132 | ||
BC119385.1 | - | 2 | 200 | 8734134 | ||
BC144982.1 | - | 2 | 200 | 9053530 |