Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bend6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177235.3 | NP_796209.2 | 3 | 360 | ||
GenBank mRNA | AK044633.1 | - | 2 | 299 | ||
AK045534.1 | - | 2 | 498 | |||
AK134655.1 | - | 3 | 502 | |||
BC057378.1 | - | 3 | 360 | 5707039 |