Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177242.4 | NP_796216.2 | 5 | 1092 | ||
GenBank mRNA | AK078914.1 | - | 5 | 1100 | ||
AK083481.1 | - | 5 | 1109 | |||
AK135118.1 | - | 5 | 1126 | |||
AK162261.1 | - | 5 | 656 | |||
BC038622.1 | - | 5 | 732 | |||
BC068149.1 | - | 5 | 1092 | 30537082 |