Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315532.1 | NP_001302461.1 | 2, 3 | 226 | ||
NM_002854.2 | NP_002845.1 | 3, 4 | 246 | |||
GenBank mRNA | AK311832.1 | - | 2, 3 | 226 | ||
AV724364.1 | - | 2, 3 | 205 | |||
BC069300.1 | - | 3, 4 | 246 | 7262744 | ||
BC096112.3 | - | 3, 4 | 252 | 40002305 | ||
BC096113.3 | - | 3, 4 | 252 | 40002307 | ||
BC096114.1 | - | 3, 4 | 252 | 40002308 | ||
BC096115.1 | - | 3, 4 | 252 | 40002310 | ||
CR456552.1 | - | 2, 3 | 212 | |||
DA054941.1 | - | 2, 3 | 227 | |||
DA392417.1 | - | 2, 3 | 226 | |||
X63070.1 | - | 3, 4 | 252 |