Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001798.2 | NP_001001798.1 | 18 | 1998 | ||
NM_001037863.1 | NP_001032952.1 | 18 | 1998 | |||
GenBank mRNA | AK039243.1 | - | 18 | 1999 | ||
AK043065.1 | - | 18 | 1992 | |||
AK143860.1 | - | 17 | 1876 | |||
BC106087.1 | - | 18 | 1977 | 30843359 |