Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159389.1 | NP_001152861.1 | 15 | 1629 | ||
NM_177306.3 | NP_796280.1 | 12 | 1260 | |||
GenBank mRNA | AK029809.1 | - | 12 | 1261 | ||
BC132104.1 | - | 10 | 943 | 40130381 | ||
BC132294.1 | - | 10 | 943 | 40130571 | ||
BC145065.1 | - | 9 | 825 | |||
FJ804414.1 | - | 14 | 1477 |