Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111017.1 | NP_001104487.1 | 9 | 896 | ||
NM_177311.4 | NP_796285.2 | 8 | 806 | |||
GenBank mRNA | AK019677.1 | - | 8 | 795 | ||
AK154605.1 | - | 9 | 847 | |||
AK155571.1 | - | 9 | 899 | |||
AK162246.1 | - | 9 | 831 | |||
AK170498.1 | - | 9 | 847 | |||
BC119594.1 | - | 9 | 885 | 40059066 | ||
BC119595.1 | - | 9 | 885 | 40059071 |