Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001491.1 | NP_001001491.1 | 7 | 683 | ||
GenBank mRNA | AK206089.1 | - | 4 | 218 | ||
AK207394.1 | - | 4 | 216 | |||
BC023701.1 | - | 7 | 690 | 5345587 | ||
BC023827.1 | - | 7 | 677 | 5343662 | ||
BC032174.1 | - | 7 | 679 | |||
BC032175.1 | - | 7 | 679 | |||
BC070421.1 | - | 7 | 683 | 6403300 |