Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037713.4 | NP_001032802.2 | 4 | 456 | ||
GenBank mRNA | AK046736.1 | - | 3 | 227 | ||
AK140315.1 | - | 4 | 421 | |||
CK130151.1 | - | 3 | 391 |