Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183201.4 | NP_899024.3 | 3 | 1068 | ||
GenBank mRNA | AK036486.1 | - | 3 | 1069 | ||
AK040359.1 | - | 3 | 1055 | |||
AK171886.1 | - | 2 | 1010 | |||
AY261804.1 | - | 2 | 1030 | |||
AY261805.1 | - | 2 | 1025 | |||
BC117909.1 | - | 2 | 948 | 40097236 | ||
BC117910.1 | - | 2 | 948 | 40097237 |