Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144953.1 | NP_001138425.1 | 22 | 3604 | ||
GenBank mRNA | AK045595.1 | - | 1 | 1594 | ||
BC110635.2 | - | 1 | 1333 | |||
BC125016.1 | - | 1 | 1335 | |||
BE862588.1 | - | 3 | 252 |